Anti-ACHE Mouse Monoclonal Antibody [clone: 684CT8.3.4]

Supplier: Abgent
AM2184B
10217-850EA 601.98 USD
10217-850
Anti-ACHE Mouse Monoclonal Antibody [clone: 684CT8.3.4]
Antibodies
Western Blot: 1:2000

Type: Primary
Antigen: ACHE
Clonality: Monoclonal
Clone: 684CT8.3.4
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR