Anti-TFAP2C Rabbit Polyclonal Antibody

Supplier: Abgent
AP20531A
10217-820EA 589.62 USD
10217-820
Anti-TFAP2C Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: TFAP2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR