Anti-GNB1A Rabbit Polyclonal Antibody

Supplier: Abgent
AZB10033A
10217-800EA 589.62 USD
10217-800
Anti-GNB1A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: GNB1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR