Anti-FUBP3 Mouse Monoclonal Antibody [clone: 1216CT820.155.150]

Supplier: Abgent
AM2236B
10217-794EA 586.14 USD
10217-794
Anti-FUBP3 Mouse Monoclonal Antibody [clone: 1216CT820.155.150]
Antibodies
Western Blot: 1:2000, Immunofluorescence: 1:25,

Type: Primary
Antigen: FUBP3
Clonality: Monoclonal
Clone: 1216CT820.155.150
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR