Anti-ACE2 Mouse Monoclonal Antibody [clone: 881CT16.4.4]

Supplier: Abgent
AM2197A
10217-782EA 586.14 USD
10217-782
Anti-ACE2 Mouse Monoclonal Antibody [clone: 881CT16.4.4]
Antibodies
Western Blot: 1:2000

Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: 881CT16.4.4
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR