Anti-MGMT Mouse Monoclonal Antibody [clone: 888CT22.2.4]

Supplier: Abgent
AM2217A
10217-774EA 558.24 USD
10217-774
Anti-MGMT Mouse Monoclonal Antibody [clone: 888CT22.2.4]
Antibodies
Western Blot: 1:2000

Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: 888CT22.2.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR