Anti-BLMH Rabbit Polyclonal Antibody

Supplier: Abgent
AP20513C
10217-760EA 589.3 USD
10217-760
Anti-BLMH Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25,

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR