Anti-TIMM9 Rabbit Polyclonal Antibody

Supplier: Abgent
AP17579C
10217-600EA 589.3 USD
10217-600
Anti-TIMM9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: TIMM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR