Quantofix Lubricheck Test Strips, CTL Scientific Supply
Supplier: Macherey-Nagel
91336
10217-568EA 69.71 USD
10217-568
Quantofix Lubricheck Test Strips, CTL Scientific Supply
Test Paper Indicator Paper
Quantofix Lubricheck test strip for the semi-quantitative determination of the concentration of cooling lubricants
Quantofix Lubricheck has a gradation of scale: 0-15-50-75-130-200 mmol/L KOH.
The easy dip and read procedure provides a reliable result within 1 minute.
Ordering information: Packaged as a box of 100.