Quantofix EDTA Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91335
10217-566EA 60.49 USD
10217-566
Quantofix EDTA Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix EDTA test strips is for the rapid and reliable determination of EDTA, NTA and other complexing agents in solutions
Quantofix EDTA has a gradation of scale: 0-100-200-300-400 mg/L EDTA.
The easy dip and read procedure provides a reliable result within 15 seconds.
Ordering information: Packaged as a box of 100.