Quantofix Chlorine Sensitive Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91339
10217-542EA 54.75 USD
10217-542
Quantofix Chlorine Sensitive Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Chlorine Sensitive test strips for the semi-quantitative determination of total chlorine.
Quantofix Chlorine Sensitive has a gradation of scale: 0-0.1-0.5-1-3-10 mg/L
Ordering information: Packaged as a box of 100.