Quantofix Peroxide Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91312
91319
91333
10217-536EA
58.96
USD
10217-536
10217-538
10217-540
Quantofix Peroxide Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Peroxide for the semi-quantitative determination of hydrogen peroxide (H2O2) and peroxides in solutions.
Color changes with H2O2 from white to blue.
Ordering information: Packaged as a box of 100.