Quantofix Cobalt Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91303
10217-510EA 71.5 USD
10217-510
Quantofix Cobalt Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Cobalt Test strips for the semi-quantitative determination of cobalt in solutions.
Quantofix Cobalt Test strips has a gradation of scale: 0-10-25-50-100-250-500-1000 mg/L.
Ordering information: Packaged as a box of 100.