Quantofix Active Oxygen Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91349
10217-500EA 71.52 USD
10217-500
Quantofix Active Oxygen Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Active Oxygen Test trips for the semi-quantitative determination of active oxygen.
Quantofix Active Oxygen Test strips have a gradation of scale: 0-4-8-15-25 mg/L PMPS (potassium monopersulphate).
Ordering information: Packaged as a box of 100.