Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply
Supplier: CTL SCIENTIFIC SUPPLY CORP
91314
10217-492EA 68.75 USD
10217-492
Quantofix Ascorbic Acid Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Ascorbic Acid Test strips for the semi-quantitative determination for vitamin C in solutions.
Quantofix Ascorbic Acid Test strips have a gradation of scale: 0-50-100-200-300-500-1000-2000 mg/L Vitamin C. Store test strips in a cool and dry place, protected from sunlight and moisture.
Ordering information: Packaged as a box of 100.