Safe-it®, Tamper Proof Labels, Bürkle
Supplier: BURKLE INC
5303-5000
10217-336PK 202.44 USD
10217-336
Safe-it®, Tamper Proof Labels, Bürkle
Labels
Safe-it® labels are used as protection from manipulations of samples, retention samples, for transporting samples, sealing containers with important or dangerous chemicals in the lab as well as sealing letters, shipments and more.
  • With checkerboard pattern
  • Visible proof of tampering
When the safe-it security seal has been broken, a silver checkerboard pattern appears on both the subsurface and surface of the sticker itself.