Scoop, Stainless Steel, Bürkle
Supplier: BURKLE INC
5354-1000
10217-178EA 712.14 USD
10217-178
Scoop, Stainless Steel, Bürkle
Scoops
The stainless steel scoop has no openings, undercuts or crevices in which residues could collect
Pore-free finishing produced by high-quality laser welding seams. The stainless steel scoop can be flame-sterilised and can therefore be made completely germ-free.