ChemoSampler, Bürkle
Supplier: BURKLE INC
5336-1000
10217-152EA 219.63 USD
10217-152
ChemoSampler, Bürkle
Samplers Fluid Samplers
Specially developed for sample collection of aggressive liquids such as acids, alkalis, cleaning agents, (please check chemical resistance).
  • Resistant to acid and alkalis
  • Two bottle sizes available
The sample is directly extracted into the original sample bottle with screw cap. The bottle closure with tamper-evident safety ring ensures reliable sample transfer. The extension rod makes it possible to extract samples to a depth of 2000 mm, e.g. from tanks, silos or tankers. Basic length 1000 mm, extension rod 1000 mm.