UniSampler® 'Ex', Sampler, Bürkle
Supplier: BURKLE INC
5314-3000
10217-066EA 893.87 USD
10217-066
UniSampler® 'Ex', Sampler, Bürkle
Samplers Fluid Samplers
Sampler especially developed for sampling flammable liquids of Hazard Category AI (VbF, e.g. fuel).
  • Sampler for liquid samples from precisely pre-determined depth, with hand-operated vacuum pump
  • PTB approval D- 333-009 618/00
Glass bottle and hose are connected in such a way that they conduct electricity and are grounded via a grounding cable. Operation: Ground the device, immerse the suction hose to the required depth, attach the adapter to the sampling bottle, generate a vacuum with the manual pump and collect the sample, remove the filled sampling bottle, close it and label it.
Ordering information: Supplied complete with handy transport case, 4 m hose (NBR), glass bottle 1000 ml, bottle adapter, bottle stand, vacuum pump, earthing cable and hose-weight made of brass.