UniSampler®, Sampler with Hose, Bürkle
Supplier: BURKLE INC
5314-2000
10217-062EA 504.65 USD
10217-062
UniSampler®, Sampler with Hose, Bürkle
Samplers Fluid Samplers
Hand operated vacuum pump sampler, equipped with a 2.5 metre long PVC hose. Ideal for deep, narrow and hard to reach spots.
  • Powerful suction for viscous media
  • Easy to disassemble and clean
The stainless steel weight will sink the end of the hose to the required sampling depth. Liquid only comes into contact with the suction hose and sample bottle
Universal bottle adapter made of PVC, for bottle necks dia. 24 mm, 32 mm, 39 mm and 50 mm nominal dimension.