Anti-RNA Helicase A Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-28915
102162-826EA 564.74 USD
102162-826
Anti-RNA Helicase A Goat Polyclonal Antibody
Antibodies
The RNA Helicase A Antibody from Novus Biologicals is a goat polyclonal antibody to RNA Helicase A. This antibody reacts with human. The RNA Helicase A Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: RNA Helicase A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR