LaboPlast® Stopcock PA, Electroconductive, Bürkle
Supplier: BURKLE INC
0525-1100
10216-912EA 65.59 USD
10216-912
LaboPlast® Stopcock PA, Electroconductive, Bürkle
Stopcocks
Stopcock with tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm.
  • With discharge angle and rotatable spout preventing drips
  • Media-contacting parts PA conductive to electricity and PTFE
  • Supplied with PA adapter conductive to electricity for 3/4" inner thread
Stopcock PA made of electrically conductive polyamide, suitable for solvents.