Anti-DNA/RNA Damage Mouse Monoclonal Antibody [clone: 15A3]

Supplier: Novus Biologicals
NB110-96878
102126-460EA 677.97 USD
102126-460
Anti-DNA/RNA Damage Mouse Monoclonal Antibody [clone: 15A3]
Antibodies
The DNA / RNA Damage Antibody (15A3) from Novus Biologicals is a mouse monoclonal antibody to DNA / RNA Damage. This antibody reacts with non-species specific. The DNA / RNA Damage Antibody (15A3) has been validated for the following applications: ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot.

Type: Primary
Antigen: DNA/RNA Damage
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR