Anti-DNA Ligase IV Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-57379
102123-314EA 564.74 USD
102123-314
Anti-DNA Ligase IV Rabbit Polyclonal Antibody
Antibodies
The DNA Ligase IV Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNA Ligase IV. This antibody reacts with human. The DNA Ligase IV Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: DNA Ligase IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR