Anti-RNA Helicase A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40579
102121-920EA 649.66 USD
102121-920
Anti-RNA Helicase A Rabbit Polyclonal Antibody
Antibodies
The RNA Helicase A Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNA Helicase A. This antibody reacts with human, mouse. The RNA Helicase A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RNA Helicase A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: The epitope recognized by NB 110-40578 maps to a region between residue 1220 and the C-terminus (residue 1270) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 9 using the numbering given in entry CAA71668.1 (GeneID 1660).
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR