Anti-DNA Ligase III Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-81661
102118-494EA 635.51 USD
102118-494
Anti-DNA Ligase III Rabbit Polyclonal Antibody
Antibodies
The DNA Ligase III Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNA Ligase III. This antibody reacts with human. The DNA Ligase III Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: DNA Ligase III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR