Anti-RNase L Mouse Monoclonal Antibody (DyLight 488) [clone: 2E9]

Supplier: Novus Biologicals
NB100-351G
102108-578EA 626.46 USD
102108-578
Anti-RNase L Mouse Monoclonal Antibody (DyLight 488) [clone: 2E9]
Antibodies
The RNase L Antibody (2E9) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to RNase L. This antibody reacts with human, hamster, primate, mouse (negative), rat (negative). The RNase L Antibody (2E9) [DyLight 488] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RNase L
Clonality: Monoclonal
Clone: 2E9
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Hamster, Primate, Mouse (Negative), Rat (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR