Paraffin Wax Dispenser, Boekel Scientific
Supplier: Boekel
145600
10210-954EA 2232.65 USD
10210-954
Paraffin Wax Dispenser, Boekel Scientific
Paraffin Wax Trimmers and Dispensers
The Boekel Scientific Paraffin Wax Dispenser features rapid warm up and excellent thermal transfer.
  • 5 Liter (1.25 Gallon) Capacity
  • Advanced Safety Over Temp Cut Off
  • Stainless Steel Outer Casing
  • Precise Control of Wax with a Locking Dispenser Valve
  • Rapid Warm Up
The heated valve ensures consistent flow of wax while the inner reservoir screen helps to prevent large particulate from clogging the dispenser valve.
Certifications: UL/CSA and CE Marked