FREE-SAT NT1-912 Cobalt Blue, Sterile, Pre-Saturated Poly-Cellulose Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
FS-NT1-912CB
10210-953CS 455.91 USD
10210-953
FREE-SAT NT1-912 Cobalt Blue, Sterile, Pre-Saturated Poly-Cellulose Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These 9×12" wipes are pre-saturated with 70% Isopropyl Alcohol and 30% DI water and are packaged in convenient re-sealable bags.
  • Cobalt-60 gamma irradiated
  • Each packaged identified with lot number for traceability
  • Each case includes Certificate of Sterility
STERILE COBALT BLUE FREE-SAT NT1-912 Presaturated wipes are made of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform tough fabric.
Certifications: Validated to SAL (Sterility Assurance Level) 10⁻⁶ in accordance with ANSI/AAMI/ISO 11137:2006 guidelines. ISO Class 5.Meets and exceeds USP 797 and USP 800 requirements for critical contamination control