Corning® Accutase® Cell Detachment Solution, Corning
Supplier: Corning
25-058-CI
10210-214EA 63.1 USD
10210-214
Corning® Accutase® Cell Detachment Solution, Corning
Cell Separation Media
Accutase® is an effective cell detachment solution for the routine detachment of cells from standard tissue culture plasticware as well as enhanced surface and advanced surface coated cultureware
  • Efficient, with gentle detachment of adherent cells within minutes
  • An alternative to Trypsin
  • Able to eliminate neutralization or wash steps
  • Convenient and cost effective
Accutase® is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting. It contains proprietary marine-origin enzymes with proteolytic and collagenolytic activity for the detachment of primary and stem cell lines and tissues. Accutase® provides a reduced contamination risk, because it is free of mammalian or bacteria-derived products.