EMS 9002 Vacuum Processor, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
97050-A
102099-930EA 3464.84 USD
102099-930
EMS 9002 Vacuum Processor, Electron Microscopy Sciences
Tissue Processors
Made from Pyrex Glass, which offers better temperature conductivity and stability than any other material and is chemically resistant and microwave transparent.
The EMS 9002 when used in conjunction with our EMS 9000 processor improves ultra structural preservation during microwave assisted chemical fixation and reduces fixation and infiltration times dramatically.
Accessories information: The vacuum processor kit includes vacuum processor bowl, processor tray, vacuum processor tubing kit, and 74 cassette processing rack for processing of tissue samples in the microwave processor.