Gold, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-AU 91007-AU 91008-AU 91009-AU 91010-AU 91013-AU 91014-AU 91015-AU 91016-AU 91017-AU 91018-AU 91019-AU 91020-AU
102099-398EA 647.62 USD
102099-398 102099-428 102099-458 102099-488 102099-516 102099-548 102099-578 102099-608 102099-638 102099-668 102099-698 102099-728 102099-758
Gold, sputtering target sputter coater cathodes
Gold

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Au
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR