Edwards RV3 Vacuum Pump, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
91003 91003-E
102091-394EA 8097.68 USD
102091-394 102099-392
Edwards RV3 Vacuum Pump, Electron Microscopy Sciences
Pumps Vacuum Pumps
In high vacuum mode it is ideal for backing turbo pumps, analytical instruments, and electron microscopes. Configurable for high throughput mode. Ultra Grade 19 oil is recommended for use with the RV3.
  • Speed - (Pneurop 6602)
  • 50 Hz operation - 2 ft 3min-1 (3.3 m3h-1)
  • 60 Hz operation - 2.3 ft 3min-1 (3.9 m 3h-1)
  • Ultimate Vacuum (Total Pressure) - High Vacuum Mode - 2 x 10-3 mbar
  • Inlet connection - NW25