Rocker Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77512-20
102098-470RL 46.72 USD
102098-470 102098-472
Rocker Tape, Electron Microscopy Sciences
Tapes
Convenient - No more fumbling with clamps
Double-sided adhesive film for laboratory container mounting.
Convenient - No more fumbling with clamps
Versatile - Use with rockers, shakers, incubators or nutators
Strong - Easily rock and roll up to 300 rpm and beyond*
Long Lasting - Re-usable based on load capacity and vessel number
Resistent - Keeps on rockin’ even in the prescence of most solvents and caustic agents
* RPM limits may vary based on load and vessel contact area. Test load recommended
* RPM limits may vary based on load and vessel contact area. Test load recommended