Set Screw Vise, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75941-01
102098-260EA 43.97 USD
102098-260
Set Screw Vise, Electron Microscopy Sciences
Specimen Mounting Supplies
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 4 mm wide x 5 mm deep. And has a 3.2 mm dia. pin.