QuadMag Genie®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72402-12
102098-014EA 1322.56 USD
102098-014 102098-016
QuadMag Genie®, Electron Microscopy Sciences
Stirrers Magnetic Stirrers, Multi-position
QuadMag Genie® is a general purpose multi-position magnetic stirrer designed for general laboratory applications which provides vigorous stirring for up to 4 vessels simultaneously.
The powerful magnet and motor offer exceptional magnetic force capable of stirring up to 2 L for each position. Powerful 'rare earth' magnets automatically pull stir bars into position and stay coupled even under the most demanding stirring conditions. Rugged ABS/Polycarbonate housing and the famous Vortex-Genie® heavy-duty motor provide years of reliable service. Spill-resistant housing channels fluids away from the internal components.