Slide Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71411-01
102097-508EA 73.85 USD
102097-508
Slide Holder, Electron Microscopy Sciences
Slide Holders
Molded from polypropylene. Adjustable to hold 2 to 5 3x1” slides, or 10 slides back-to-back firmly. This Slide Holder works well with all Neat Stain Kits.