Slide Warmers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71319-10 71319-250 71319-50 71320-10
102097-468EA 573.9 USD
102097-468 102097-472 102097-474 102097-476
Slide Warmers, Electron Microscopy Sciences
Slide Warmers/Dryers
Step Up™ Slide Warmer’s unique patent pending design helps you save space while still meeting a busy lab’s needs
The anodized black surface provides contrast with samples. The trays are removable for easy cleaning. It has digital LED temperature display and temperature setting.
The Step Up™ holds twice as many slides as a flat unit of the same size and also makes retrieving your slides easier since the end of the slide is not on the flat surface. Model XH-2003.