Parafilm Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70993-01 70993-02 70993-03 70993-04
102097-218EA 96.16 USD
102097-218 102097-220 102097-222 102097-224
Parafilm Dispenser, Electron Microscopy Sciences
Sealing Film Dispensers
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company