Disposable Delta Spreaders, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70498-40
102097-026CS 174.05 USD
102097-026
Disposable Delta Spreaders, Electron Microscopy Sciences
Spreaders Cell Spreaders
Smooth spreading surface achieved with manufacturing parting line near top of head.
  • Edgeless Design - Soft, rounded corners won't tear agar.
  • Even Pressure - Unique delta shape smoothly distributes media.
  • Ripple Handle - Allows for easy and comfortable manipulation.
  • Sterile - May be used right out of the box.
  • Measurements - 172mm x 37mm, 4mm diameter.