Sliding Guide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70494-30
102096-958EA 296.16 USD
102096-958 102096-960
Sliding Guide, Electron Microscopy Sciences
Well Plate Stands and Well Orienters
Easy microplate viewing for pipetting and liquid transfer.
  • Easily align a pipette with microplate wells using this Sliding Guide
  • Sliding Guide is clearly marked with row letters for fast and easy orientation
  • Reduce eyestrain
  • Adjust the guide height to accommodate flat or v-bottom microplates or PCR plates
Compatible with multi-channel and single-channel pipettes.