Wash-N-Dry™ Slide Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70366-17
102096-892EA 84.43 USD
102096-892
Wash-N-Dry™ Slide Rack, Electron Microscopy Sciences
Slide Holders
Holds 12 Slides Securely holds slides for convenient cleaning, treating, staining, and drying.
Maximum Circulation Offset design allows liquid and air to completely flow around the entire slide ensuring cleanliness and faster drying times. "Hands Off Design" Tailored to contact a minimal amount of slide surface area permitting solutions and air to affect entire slide. Fits standard slide sizes including 25 mm and 1". Laboratory Friendly Utilizes chemically and thermally resistant glass infused polypropylene. Autoclavable. Create A System Use in conjunction with a 200 mL tall form beaker to efficiently use solutions.