Germinator 220 V, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66118-20
102095-946EA 920.44 USD
102095-946
Germinator 220 V, Electron Microscopy Sciences
Sterilizers Glass Bead Sterilizers
The GERMINATOR 500™ dry sterilizer is a convenient effective way to quickly decontaminate your micro-dissecting instruments. It has an unique stainless steel glass bead bath that remains at a constant 500° F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them.
  • 220 V model
  • Easy to use
  • No more need for messy chemicals or open flames in the lab
  • The small footprint saves bench space
  • Fast, decontaminates most micro-dissecting instruments within 15 seconds
  • Effective decontamination
  • Proven durable design with 1 year
More Product Information