Germinator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66118-10
102095-944EA 839.7 USD
102095-944
Germinator, Electron Microscopy Sciences
Sterilizers Glass Bead Sterilizers
The GERMINATOR 500™ dry sterilizer is a convenient effective way to quickly decontaminate your micro-dissecting instruments. It has an unique stainless steel glass bead bath that remains at a constant 500° F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them.
  • Easy to use
  • No more need for messy chemicals or open flames in the lab
  • The small footprint saves bench space
  • Fast, decontaminates most micro-dissecting instruments within 15 seconds
  • Effective decontamination
  • Proven durable design with 1 year warranty
  • Modular cord
The unit measures 6.75×5.25×5.25" (H×W×D) and the Stainless Steel Well measures: 2.0 diameter × 4.0" deep (ID). The unit only weighs 4.5 lbs.
Certifications: UL listed.