Overhead Stirrer Complete Unit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64806-10
102095-710EA 2758.62 USD
102095-710
Overhead Stirrer Complete Unit, Electron Microscopy Sciences
Stirrers Overhead Stirrers
The Wheaton overhead stirrer is a compact unit with solid state speed control capable of providing RPM/torque combinations for most laboratory operation such as homogenizing or mixing.
  • Stirrer Unit allows for Power Homogenization or Mixing
  • Use only with Tissue Grinders with Stainless Steel Rods
  • Keyless Chuck accepts Shaft Size to 7.9 mm
  • One-Year Warranty
This Overhead mixer should be attached only to tissue grinders with Teflon® pestles and with stainless steel rods. Unit and motor are mounted in a housing with epoxy finish, and it is provided with a 6" aluminum rod and clamp for attachment to support stand. Support stand is not included.