Micro Tissue Grinder Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64800 64799-02
102091-106EA 1248.5 USD
102091-106 102095-700
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Homogenizers Tissue Grinders
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
  • Complete kit includes 7 micro tissue grinders
  • Made from Wheaton 33 low extractable borosilicate glass
  • Replacement parts available separately
  • Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.