Advanced Protocol Microbiological Incubators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63154-20
102094-512EA 8041.21 USD
102094-512
Advanced Protocol Microbiological Incubators, Electron Microscopy Sciences
Incubators Standard Incubators
Dual convection for application versatility-fan speed adjustable from 0 to 100%
  • Intuitive user interface for setting temperature
  • Large, easy to read vacuum fluorescent display
  • Internal glass door allows sample viewing without impacting temperature
  • Exceptionally small footprint
  • Easy to clean, corrosion-resistant stainless-steel interior (AISI 430)
  • Advanced digital timer for daily or weekly ON / OFF cycles
The gravity convection incubator provides temperatures at or below ambient: +5 to 105°C. It has a temperature uniformity of ±0.6 °C (at 37 °C) and temperature stability of ± 0.1 °C (at 37 °C).