Simport MEGA Double Cytology Funnels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62754-10
102094-450PK 181 USD
102094-450
Simport MEGA Double Cytology Funnels, Electron Microscopy Sciences
Funnels Cytology Funnels
This ALL PLASTIC CytoSep™ MEGA Funnel provides an easy, efficient and cost effective method of producing high quality thin-layer slide preparations. It eliminates the needs for stainless steel clips. It is ideally for preparation of large volume samples. The larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22 x 14.75 mm (325 mm squared) provides for up to 12 times the sample volume (6 ml) of the single Cyto™ Cytology Funnel. Strategically placed baffles inhibit cell setting which results in uniform cell deposition and excellent quality slide preparations. This MEGA funnels and cap minimize user exposure to pathogens while reducing the risk of specimen cross contamination. Can be used wtih both air dried and fixed preparations.