True North™ Cool Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62013-04 62013-10 62013-40 62013-90
102093-894EA 159.15 USD
102093-894 102093-896 102093-898 102093-900
True North™ Cool Containers, Electron Microscopy Sciences
Boxes Cryoboxes
These rigid single mold polyurethane containers provides durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra colduse with ice, LN2, ice-salt slurries, and more.
  • Convenient lid nests beneath bucket or pan and easily removes with comfortable recessed knobs
  • Snug fit minimizes evaporation loss
  • Handy pour spout for pouring melted ice, water, and solutions
  • Ergonomic handles safely transport bucket or pan from work area to sink
  • Deep internal cavity has enough room to add a rack
  • Tapered sides make stacking and storage compact
  • Available in a bucket or pan and in multiple sizes