CoolSafe Cooling Chambers Individual Components, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61951-10 61951-20 61951-30 61951-40 61951-50 61951-60 61951-80 61951-70
102093-826EA 596.85 USD
102093-826 102093-828 102093-830 102093-832 102093-834 102093-836 102093-840 102681-020
CoolSafe Cooling Chambers Individual Components, Electron Microscopy Sciences
Coolers Benchtop Coolers
Each cooling chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.